siRNA Details
Gene Symbol: Olr1331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000790.1 | NP_001000790.1 | 585 | |||
XM_008766124.1 | XP_008764346.1 | 2 | 474 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000790.1 | NP_001000790.1 | 585 | |||
XM_008766124.1 | XP_008764346.1 | 2 | 474 |