siRNA Details
| Gene Symbol: Olr1316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000793.1 | NP_001000793.1 | 1 | 517 | ||
| Gene Symbol: Olr1319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000470.1 | NP_001000470.1 | 1 | 517 | ||

Olr1316
Genomic Map