siRNA Details
| Gene Symbol: Olr1262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000803.1 | NP_001000803.1 | 1 | 584 | ||
| Gene Symbol: LOC102552923 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235498.1 | XP_006235560.1 | 1 | 584 | ||
| Gene Symbol: Olr1261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000804.1 | NP_001000804.1 | 1 | 584 | ||

Olr1262
Genomic Map