siRNA Details
| Gene Symbol: Olr1194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000820.1 | NP_001000820.1 | 1 | 490 | ||
| Gene Symbol: Olr1195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001081.1 | NP_001001081.1 | 1 | 490 | ||

Olr1194
Genomic Map