siRNA Details
| Gene Symbol: Olr407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000828.1 | NP_001000828.1 | 1 | 237 | ||
| Gene Symbol: Olr406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000383.1 | NP_001000383.1 | 1 | 237 | ||

Olr407
Genomic Map