siRNA Details
| Gene Symbol: Olr1633 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000835.1 | NP_001000835.1 | 1 | 257 | ||
| Gene Symbol: LOC103690304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770628.1 | XP_008768850.1 | 1 | 289 | ||
| Gene Symbol: Olr1635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000970.1 | NP_001000970.1 | 302 | |||
More Information
Set Membership: |
|

Olr1633
Genomic Map