siRNA Details
Gene Symbol: Olr1631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000837.1 | NP_001000837.1 | 1 | 455 |
Gene Symbol: Olr1633 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000835.1 | NP_001000835.1 | 1 | 422 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
