siRNA Details
| Gene Symbol: Olr821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000842.1 | NP_001000842.1 | 1 | 910 | ||
| Gene Symbol: Olr820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000974.1 | NP_001000974.1 | 1 | 910 | ||

Olr821
Genomic Map