siRNA Details
| Gene Symbol: Olr803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000853.1 | NP_001000853.1 | 1 | 451 | ||
| Gene Symbol: Olr801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001004.1 | NP_001001004.1 | 1 | 454 | ||
| Gene Symbol: Olr802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000977.1 | NP_001000977.1 | 1 | 454 | ||

Olr803
Genomic Map