siRNA Details
| Gene Symbol: KCNQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004700.3 | NP_004691.2 | 5, 6 | 923 | ||
| NM_172163.2 | NP_751895.1 | 5, 6 | 923 | |||
| XM_017002792.1 | XP_016858281.1 | 2, 3 | 164 | |||
| GenBank mRNA | AF105202.1 | - | 5, 6 | 923 | ||
| AK074957.1 | - | 2, 3 | 160 | |||
| DA782056.1 | - | 2, 3 | 215 | |||

KCNQ4
Genomic Map