siRNA Details
| Gene Symbol: Olr1162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000870.1 | NP_001000870.1 | 1 | 266 | ||
| Gene Symbol: Olr1160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000985.1 | NP_001000985.1 | 1 | 266 | ||

Olr1162
Genomic Map