siRNA Details
| Gene Symbol: Olr1710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000886.1 | NP_001000886.1 | 1 | 595 | ||
| Gene Symbol: Olr1708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000276.1 | NP_001000276.1 | 1 | 571 | ||

Olr1710
Genomic Map