siRNA Details
| Gene Symbol: Olr1689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000889.1 | NP_001000889.1 | 1 | 563 | ||
| Gene Symbol: LOC100910420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751906.2 | XP_003751954.1 | 580 | |||

Olr1689
Genomic Map