siRNA Details
| Gene Symbol: Olr1682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000890.1 | NP_001000890.1 | 1 | 78 | ||
| Gene Symbol: Olr1681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001006.1 | NP_001001006.1 | 1 | 78 | ||

Olr1682
Genomic Map