siRNA Details
| Gene Symbol: Olr1680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000891.1 | NP_001000891.1 | 1 | 545 | ||
| Gene Symbol: LOC100912354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751901.3 | XP_003751949.3 | 1 | 146 | ||

Olr1680
Genomic Map