siRNA Details
| Gene Symbol: Olr837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000897.1 | NP_001000897.1 | 1 | 640 | ||
| GenBank mRNA | AF091565.1 | - | 1 | 448 | ||
| Gene Symbol: LOC103692344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763684.1 | XP_008761906.1 | 1033 | |||

Olr837
Genomic Map