siRNA Details
| Gene Symbol: Olr832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000898.1 | NP_001000898.1 | 1 | 643 | ||
| Gene Symbol: Olr831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000899.1 | NP_001000899.1 | 1 | 643 | ||

Olr832
Genomic Map