siRNA Details
| Gene Symbol: Olr1598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000910.1 | NP_001000910.1 | 1 | 52 | ||
| Gene Symbol: Olr1600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000086.1 | NP_001000086.1 | 1 | 49 | ||
More Information
Set Membership: |
|

Olr1598
Genomic Map