siRNA Details
| Gene Symbol: Olr611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000926.1 | NP_001000926.1 | 412 | |||
| Gene Symbol: Olr610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000336.1 | NP_001000336.1 | 1 | 412 | ||

Olr611
Genomic Map