siRNA Details
| Gene Symbol: LOC103690410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759846.1 | XP_008758068.1 | 1 | 561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103690410| Gene Symbol: LOC103690410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759846.1 | XP_008758068.1 | 1 | 561 | ||