siRNA Details
| Gene Symbol: Olr194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000948.1 | NP_001000948.1 | 1 | 754 | ||
| Gene Symbol: Olr196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000187.1 | NP_001000187.1 | 1 | 754 | ||

Olr194
Genomic Map