siRNA Details
| Gene Symbol: Olr1451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000957.1 | NP_001000957.1 | 853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1451| Gene Symbol: Olr1451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000957.1 | NP_001000957.1 | 853 | |||