siRNA Details
| Gene Symbol: Olr1279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000961.1 | NP_001000961.1 | 1 | 809 | ||
| Gene Symbol: LOC100910543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231901.1 | XP_006231963.1 | 1 | 809 | ||
| Gene Symbol: LOC103691445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760717.1 | XP_008758939.1 | 1 | 809 | ||

Olr1279
Genomic Map