siRNA Details
| Gene Symbol: Olr1590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000995.1 | NP_001000995.1 | 1 | 410 | ||
| Gene Symbol: Olr1591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000912.1 | NP_001000912.1 | 1 | 410 | ||

Olr1590
Genomic Map