siRNA Details
| Gene Symbol: Olr264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000999.1 | NP_001000999.1 | 1 | 622 | ||
| Gene Symbol: Olr267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000228.1 | NP_001000228.1 | 1 | 622 | ||

Olr264
Genomic Map