siRNA Details
| Gene Symbol: Olr1681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001006.1 | NP_001001006.1 | 1 | 676 | ||
| Gene Symbol: Olr1678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000893.1 | NP_001000893.1 | 1 | 676 | ||
| Gene Symbol: Olr1683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000269.1 | NP_001000269.1 | 1 | 676 | ||

Olr1681
Genomic Map