siRNA Details
| Gene Symbol: Olr1174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001016.1 | NP_001001016.1 | 1 | 475 | ||
| Gene Symbol: Olr1867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101022.1 | NP_001094492.1 | 1 | 550 | ||
| GenBank mRNA | D12820.1 | - | 1 | 550 | ||

Olr1174
Genomic Map