siRNA Details
| Gene Symbol: Olr129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001027.1 | NP_001001027.1 | 1 | 76 | ||
| Gene Symbol: Olr130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001272.1 | NP_001001272.1 | 1 | 76 | ||

Olr129
Genomic Map