siRNA Details
| Gene Symbol: Olr198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001033.1 | NP_001001033.1 | 1 | 587 | ||
| Gene Symbol: Olr199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001034.1 | NP_001001034.1 | 1 | 587 | ||
| Gene Symbol: Olr200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000189.1 | NP_001000189.1 | 1 | 587 | ||

Olr198
Genomic Map