siRNA Details
| Gene Symbol: Olr199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001034.1 | NP_001001034.1 | 1 | 694 | ||
| Gene Symbol: Olr198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001033.1 | NP_001001033.1 | 1 | 694 | ||

Olr199
Genomic Map