siRNA Details
| Gene Symbol: Olr232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001035.1 | NP_001001035.1 | 1 | 629 | ||
| Gene Symbol: Olr233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000207.1 | NP_001000207.1 | 1 | 629 | ||

Olr232
Genomic Map