siRNA Details
| Gene Symbol: Olr367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001047.1 | NP_001001047.1 | 1 | 511 | ||
| Gene Symbol: Olr357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000257.1 | NP_001000257.1 | 1 | 514 | ||

Olr367
Genomic Map