siRNA Details
| Gene Symbol: Olr546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001054.1 | NP_001001054.1 | 1 | 48 | ||
| Gene Symbol: LOC100910165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234489.1 | XP_006234551.1 | 48 | |||
| Gene Symbol: Olr550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000322.1 | NP_001000322.1 | 1 | 48 | ||

Olr546
Genomic Map