siRNA Details
| Gene Symbol: Olr546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001054.1 | NP_001001054.1 | 1 | 334 | ||
| Gene Symbol: Olr540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000321.1 | NP_001000321.1 | 1 | 334 | ||

Olr546
Genomic Map