siRNA Details
| Gene Symbol: Olr984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001074.1 | NP_001001074.1 | 1 | 97 | ||
| Gene Symbol: LOC102555501 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765070.1 | XP_008763292.1 | 3 | 937 | ||
| Gene Symbol: Olr894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000940.1 | NP_001000940.1 | 1 | 97 | ||

Olr984
Genomic Map