siRNA Details
| Gene Symbol: Olr1022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001075.1 | NP_001001075.1 | 1 | 801 | ||
| Gene Symbol: Olr952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000074.1 | NP_001000074.1 | 1 | 801 | ||

Olr1022
Genomic Map