siRNA Details
| Gene Symbol: Olr1138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001080.1 | NP_001001080.1 | 1 | 577 | ||
| Gene Symbol: LOC100911944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750439.1 | XP_003750487.1 | 1 | 577 | ||

Olr1138
Genomic Map