siRNA Details
| Gene Symbol: Olr1225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001083.1 | NP_001001083.1 | 1 | 235 | ||
| Gene Symbol: LOC100911999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766103.1 | XP_008764325.1 | 247 | |||

Olr1225
Genomic Map