siRNA Details
| Gene Symbol: Olr1254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001085.1 | NP_001001085.1 | 1 | 623 | ||
| Gene Symbol: LOC100912543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762453.1 | XP_008760675.1 | 1 | 623 | ||
| Gene Symbol: LOC100912580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762454.1 | XP_008760676.1 | 1 | 623 | ||
| Gene Symbol: LOC103692041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762675.1 | XP_008760897.1 | 1 | 623 | ||
| Gene Symbol: LOC103692042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762676.1 | XP_008760898.1 | 1 | 623 | ||
| Gene Symbol: Olr1253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000805.1 | NP_001000805.1 | 1 | 623 | ||

Olr1254
Genomic Map