siRNA Details
| Gene Symbol: Olr1394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001091.1 | NP_001001091.1 | 1 | 673 | ||
| Gene Symbol: Olr1395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001092.1 | NP_001001092.1 | 1 | 673 | ||

Olr1394
Genomic Map