siRNA Details
| Gene Symbol: Olr1400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001096.1 | NP_001001096.1 | 796 | |||
| Gene Symbol: Olr1399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001095.1 | NP_001001095.1 | 1 | 796 | ||

Olr1400
Genomic Map