siRNA Details
| Gene Symbol: Olr1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001097.1 | NP_001001097.1 | 1 | 151 | ||
| XM_006246511.2 | XP_006246573.1 | 2 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1462| Gene Symbol: Olr1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001097.1 | NP_001001097.1 | 1 | 151 | ||
| XM_006246511.2 | XP_006246573.1 | 2 | 255 | |||