siRNA Details
| Gene Symbol: Olr1546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001106.1 | NP_001001106.1 | 1 | 631 | ||
| Gene Symbol: Olr1545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001105.1 | NP_001001105.1 | 1 | 631 | ||
| Gene Symbol: Olr1547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001107.1 | NP_001001107.1 | 1 | 631 | ||

Olr1546
Genomic Map