siRNA Details
| Gene Symbol: Olr1697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001111.1 | NP_001001111.1 | 1 | 641 | ||
| Gene Symbol: LOC100362856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728918.3 | XP_002728964.3 | 1 | 641 | ||
| Gene Symbol: Olr1699 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001112.1 | NP_001001112.1 | 1 | 641 | ||

Olr1697
Genomic Map