siRNA Details
| Gene Symbol: Olr1697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001111.1 | NP_001001111.1 | 1 | 124 | ||
| Gene Symbol: Olr1696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206850.1 | NP_996732.1 | 1 | 124 | ||
| GenBank mRNA | AF034896.1 | - | 346 | |||

Olr1697
Genomic Map