siRNA Details
| Gene Symbol: Olr1700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001113.1 | NP_001001113.1 | 1 | 668 | ||
| Gene Symbol: LOC103689944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255858.2 | XP_006255920.1 | 1 | 506 | ||

Olr1700
Genomic Map