siRNA Details
| Gene Symbol: Olr1720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001116.1 | NP_001001116.1 | 1 | 838 | ||
| Gene Symbol: Olr1718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214460.1 | NP_999625.1 | 1 | 838 | ||

Olr1720
Genomic Map