siRNA Details
Gene Symbol: Olr1347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001121.1 | NP_001001121.1 | 1 | 343 | ||
GenBank mRNA | AF091579.1 | - | 1 | 133 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001121.1 | NP_001001121.1 | 1 | 343 | ||
GenBank mRNA | AF091579.1 | - | 1 | 133 |