siRNA Details
| Gene Symbol: Olr444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001123.1 | NP_001001123.1 | 1 | 581 | ||
| Gene Symbol: Olr443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000285.1 | NP_001000285.1 | 1 | 581 | ||

Olr444
Genomic Map