siRNA Details
| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002828.2 | NP_001002828.1 | 1, 2 | 314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbx2| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002828.2 | NP_001002828.1 | 1, 2 | 314 | ||